missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Biphenyl-4-thiocarboxamide, 97%

Product Code. 15448817
Change view
Click to view available options
Quantity:
5g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448817 5g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15448817 Supplier Alfa Aesar™ Supplier No. H52225.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 13363-50-3
Molecular Formula C13H11NS
MDL Number MFCD02179225
Quantity 5g
Synonym 1,1'-biphenyl-4-carbothioamide, 4-biphenylthioamide, biphenyl-4-carbothioic acid amide, 4-biphenylthiocarboxamide, 4-phenylthiobenzamide, 4-phenylbenzene-1-carbothioamide, biphenyl-4-carbothioamide, biphenyl-4-thiocarboxamide, acmc-20amoc
InChI Key HYDDAJTWPBWUOB-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C(=S)N
IUPAC Name 4-phenylbenzenecarbothioamide
Molecular Weight (g/mol) 213.298
PubChem CID 15785125
Formula Weight 213.3
Percent Purity 97%
Chemical Name or Material Biphenyl-4-thiocarboxamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.