missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,3-dibromopropyl) Phosphate, TRC

Product Code. 30003458
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003458 250 mg 250mg
17982865 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30003458 Supplier Toronto Research Chemicals Supplier No. TRCB426235250MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5412-25-9
Molecular Formula C6H11Br4O4P
Molecular Weight (g/mol) 497.74
Synonym 2,3-Dibromo-1-propanol 1,1’-(Hydrogen Phosphate), Bis(2,3-dibromopropyl) Hydrogen Phosphate, Bis(2,3-dibromopropyl) Phosphate, NSC 3239
IUPAC Name bis(2,3-dibromopropyl) hydrogen phosphate
SMILES C(C(CBr)Br)OP(=O)(O)OCC(CBr)Br

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C6H11Br4O4P/c7-1-5(9)3-13-15(11,12)14-4-6(10)2-8/h5-6H,1-4H2,(H,11,12)
Formula Weight 493.7128
Percent Purity >95%
Chemical Name or Material Bis(2,3-dibromopropyl) Phosphate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.