Learn More
Bis(2,5-iodomethyl)dioxane (Mixture of Diastereomers), TRC
Product Code. 30328797
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30328797 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30328797 Supplier Toronto Research Chemicals Supplier No. TRCB4487505G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Bis(2,5-iodomethyl)dioxane (Mixture of Diastereomers)
CAS 101084-46-2
Molecular Formula C6H10I2O2
Quantity 5 g
Synonym 2,5-Bis(iodomethyl)-p-dioxane
InChI Formula InChI=1S/C6H10I2O2/c7-1-5-3-10-6(2-8)4-9-5/h5-6H,1-4H2
SMILES ICC1COC(CI)CO1
IUPAC Name 2,5-bis(iodomethyl)-1,4-dioxane
Molecular Weight (g/mol) 367.95
Formula Weight 367.877