missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-butoxyethyl) 2-(3-Hydroxybutoxy)ethyl Phosphate Triester, TRC

Product Code. 30180404
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180404 1 mg 1mg
30179877 10 mg 10mg
30180164 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180404 Supplier Toronto Research Chemicals Supplier No. TRCB4152201MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1477494-87-3
Molecular Formula C18H39O8P
Molecular Weight (g/mol) 414.47
Synonym Bis(2-butoxyethyl) 2-(3-hydroxybutoxy)ethyl ester Phosphoric Acid
IUPAC Name bis(2-butoxyethyl) 2-(3-hydroxybutoxy)ethyl phosphate
SMILES CC(O)CCOCCOP(OCCOCCCC)(OCCOCCCC)=O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C18H39O8P/c1-4-6-9-21-12-15-24-27(20,25-16-13-22-10-7-5-2)26-17-14-23-11-8-18(3)19/h18-19H,4-17H2,1-3H3
Percent Purity >95%
Chemical Name or Material Bis(2-butoxyethyl) 2-(3-Hydroxybutoxy)ethyl Phosphate Triester
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.