missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-butoxyethyl) 2-Hydroxyethyl Phosphate Triester, TRC

Product Code. 30009822
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009822 25 mg 25mg
30011259 50 mg 50mg
30012646 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009822 Supplier Toronto Research Chemicals Supplier No. TRCB41519025MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1477494-86-2
Molecular Formula C14 H31 O7 P
Molecular Weight (g/mol) 342.36
Synonym Bis(2-butoxyethyl) 2-Hydroxyethyl Phosphate Triester, Bis(2-butoxyethyl) 2-Hydroxyethyl Ester Phosphoric Acid;
IUPAC Name bis(2-butoxyethyl) 2-hydroxyethyl phosphate
SMILES CCCCOCCOP(=O)(OCCO)OCCOCCCC

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C14H31O7P/c1-3-5-8-17-11-13-20-22(16,19-10-7-15)21-14-12-18-9-6-4-2/h15H,3-14H2,1-2H3
Formula Weight 342.1807
Percent Purity >95%
Chemical Name or Material Bis(2-butoxyethyl) 2-hydroxyethyl ester phosphoric acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.