missing translation for 'onlineSavingsMsg'
Learn More

Bis[(2-diphenylphosphino)phenyl] ether, 98%

Product Code. 11365005
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11365005 5 g 5g
11355005 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11365005 Supplier Thermo Scientific Alfa Aesar Supplier No. L18481.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 166330-10-5 | C36H28OP2 | 538.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 166330-10-5
Molecular Formula C36H28OP2
Molecular Weight (g/mol) 538.57
MDL Number MFCD00233863
InChI Key RYXZOQOZERSHHQ-UHFFFAOYSA-N
Synonym bis 2-diphenylphosphinophenyl ether, dpephos, oxydi-2,1-phenylene bis diphenylphosphine, oxybis 2,1-phenylene bis diphenylphosphine, bis 2-diphenylphosphino phenyl ether, 2,2'-bis diphenylphosphino diphenyl ether, 2-2-diphenylphosphanyl phenoxy phenyl diphenylphosphane
PubChem CID 4285986
IUPAC Name [2-(2-diphenylphosphanylphenoxy)phenyl]-diphenylphosphane
SMILES O(C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 184°C to 186°C
Quantity 5 g
Beilstein 7729718
Formula Weight 538.57
Percent Purity 98%
Chemical Name or Material Bis[(2-diphenylphosphino)phenyl] ether

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.