missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl) sebacate, 95%

Product Code. 11458706
Change view
Click to view available options
Quantity:
250 mL
1000 mL
5000 mL
Unit Size:
1000mL
250mL
5000mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458706 250 mL 250mL
11448706 1000 mL 1000mL
11468706 5000 mL 5000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458706 Supplier Thermo Scientific Alfa Aesar Supplier No. A16049.AK
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 122-62-3 | C26H50O4 | 426.68 g/mol

Bis(2-ethylhexyl) sebacate is used as a plasticizer. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-ethylhexyl) sebacate is used as a plasticizer. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in water (0.1 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-62-3
Molecular Formula C26H50O4
Molecular Weight (g/mol) 426.68
MDL Number MFCD00009497
InChI Key VJHINFRRDQUWOJ-UHFFFAOYNA-N
Synonym bis 2-ethylhexyl sebacate, bisoflex, bis 2-ethylhexyl decanedioate, plexol, bisoflex dos, monoplex dos, octoil s, reolube dos, staflex dos, di 2-ethylhexyl sebacate
PubChem CID 31218
IUPAC Name bis(2-ethylhexyl) decanedioate
SMILES CCCCC(CC)COC(=O)CCCCCCCCC(=O)OCC(CC)CCCC

Specifications

Density 0.914
Boiling Point 211°C to 213°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.451
Quantity 250 mL
Merck Index 14,1251
Solubility Information Soluble in water (0.1g/L at 20°C).
Formula Weight 426.68
Percent Purity 95%
Chemical Name or Material Bis(2-ethylhexyl) sebacate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.