missing translation for 'onlineSavingsMsg'
Learn More

Bis[(2-guanidino-4-thiazolyl)methyl]disulfide (>85% Purity)(Famotidine Impurity), TRC

Product Code. 30134991
Change view
Click to view available options
Quantity:
1 mg
25 mg
250 mg
Unit Size:
1mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134991 250 mg 250mg
17961245 25 mg 25mg
17900535 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134991 Supplier Toronto Research Chemicals Supplier No. TRCB439800250MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129083-44-9
Molecular Formula C10 H14 N8 S4
Molecular Weight (g/mol) 374.53
Synonym 2,2'-[Disulfanediylbis-(methylenethiazole-4,2-diyl)]diguanidine, Famotidine Imp. E (EP)
IUPAC Name 2-[4-[[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methyldisulfanyl]methyl]-1,3-thiazol-2-yl]guanidine
SMILES NC(=Nc1nc(CSSCc2csc(N=C(N)N)n2)cs1)N

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C10H14N8S4/c11-7(12)17-9-15-5(1-19-9)3-21-22-4-6-2-20-10(16-6)18-8(13)14/h1-2H,3-4H2,(H4,11,12,15,17)(H4,13,14,16,18)
Formula Weight 374.0224
Percent Purity >95%
Chemical Name or Material 2,2'-[Disulfanediylbis-(methylenethiazole-4,2-diyl)]diguanidine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.