missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-isopropylphenyl) Phenyl Phosphate, TRC

Product Code. 30011383
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011383 50 mg 50mg
30007130 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011383 Supplier Toronto Research Chemicals Supplier No. TRCB45144550MG
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69500-29-4
Molecular Formula C24H27O4P
Molecular Weight (g/mol) 410.44
Synonym Phosphoric Acid Bis[2-(1-methylethyl)phenyl] Phenyl Ester, Bis(o-isopropylphenyl) Phenyl Phosphate;
IUPAC Name phenyl bis(2-propan-2-ylphenyl) phosphate
SMILES CC(C)C1=CC=CC=C1OP(=O)(OC2=CC=CC=C2)OC3=CC=CC=C3C(C)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C24H27O4P/c1-18(2)21-14-8-10-16-23(21)27-29(25,26-20-12-6-5-7-13-20)28-24-17-11-9-15-22(24)19(3)4/h5-19H,1-4H3
Formula Weight 410.1647
Chemical Name or Material Bis(2-isopropylphenyl) Phenyl Phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.