missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-methoxyphenyl) carbonate, 98%, Thermo Scientific™

Product Code. 11373386
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11373386 25 g 25g
11363386 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11373386 Supplier Thermo Scientific Alfa Aesar Supplier No. L12825.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 553-17-3
Molecular Formula C15H14O5
Molecular Weight (g/mol) 274.272
MDL Number MFCD00025723
InChI Key ORUJFMPWKPVXLZ-UHFFFAOYSA-N
Synonym guaiacol carbonate, bis 2-methoxyphenyl carbonate, duotal, tussophob, guaiacyl carbonate, di-o-methoxyphenyl carbonate, bis-o-methoxyphenyl carbonate, carbonic acid bis 2-methoxyphenyl ester, guaiacol carbonic acid neutral ester, unii-q71xpq6r29
PubChem CID 11104
IUPAC Name bis(2-methoxyphenyl) carbonate
SMILES COC1=CC=CC=C1OC(=O)OC2=CC=CC=C2OC

Specifications

Melting Point 86°C to 88°C
Quantity 25 g
Beilstein 2472687
Formula Weight 274.28
Percent Purity 98%
Chemical Name or Material Bis(2-methoxyphenyl) carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.