Learn More
Bis(3-morpholino-5,6-dihydropyridine)diazene Apixaban, TRC
Product Code. 30009641
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009641 1 mg 1mg
30009074 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30009641 Supplier Toronto Research Chemicals Supplier No. TRCB5812551MG
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1998079-15-4
Molecular Formula C30 H34 N6 O4
Molecular Weight (g/mol) 542.629
Synonym Apixaban Impurity 16, 5-Acetyl-1-(4-methoxyphenyl)-4-(2-((4-(2-oxopiperidin-1-yl)phenyl)amino)ethyl)-1H-pyrazole-3-carboxamide
IUPAC Name 5-morpholin-4-yl-1-[4-[(E)-[4-(5-morpholin-4-yl-6-oxo-2,3-dihydropyridin-1-yl)phenyl]diazenyl]phenyl]-2,3-dihydropyridin-6-one
SMILES O=C1N(CCC=C1N2CCOCC2)c3ccc(cc3)\N=N\c4ccc(cc4)N5CCC=C(N6CCOCC6)C5=O
Specifications
Quantity 1 mg
InChI Formula InChI=1S/C30H34N6O4/c37-29-27(33-15-19-39-20-16-33)3-1-13-35(29)25-9-5-23(6-10-25)31-32-24-7-11-26(12-8-24)36-14-2-4-28(30(36)38)34-17-21-40-22-18-34/h3-12H,1-2,13-22H2/b32-31+
Formula Weight 542.264
Chemical Name or Material Bis(3-morpholino-5,6-dihydropyridine)diazene Apixaban