Learn More
Bis(4-octylphenyl)amine, TRC
Product Code. 30335158
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30335158 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30335158 Supplier Toronto Research Chemicals Supplier No. TRCB50890050MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Bis(4-octylphenyl)amine
CAS 101-67-7
Purity Grade Notes HPLC
Molecular Formula C28 H43 N
Quantity 50 mg
Synonym Vanlube-81, p, p’-dioctyldiphenylamine
InChI Formula InChI=1S/C28H43N/c1-3-5-7-9-11-13-15-25-17-21-27(22-18-25)29-28-23-19-26(20-24-28)16-14-12-10-8-6-4-2/h17-24,29H,3-16H2,1-2H3
SMILES CCCCCCCCc1ccc(Nc2ccc(CCCCCCCC)cc2)cc1
IUPAC Name 4-octyl-N-(4-octylphenyl)aniline
Molecular Weight (g/mol) 393.65
Formula Weight 393.3396
Percent Purity >95