missing translation for 'onlineSavingsMsg'
Learn More

Bis(p-aminophenyl) Ether, TRC

Product Code. 30346623
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30346623 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30346623 Supplier Toronto Research Chemicals Supplier No. TRCB405510100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Aminophenylether (4,4'-Oxydianiline)
CAS 101-80-4
Molecular Formula C12 H12 N2 O
Quantity 100 g
Synonym Aniline, 4,4'-oxydi- (6CI,8CI), 4,4'-Oxybis[benzenamine], 1-Amino-4-(4-aminophenoxy)benzene, 4,4'-Diaminobiphenyl ether, 4,4'-Diaminobiphenyl oxide, 4,4'-Diaminodiphenyl ether, 4,4'-Diaminodiphenyl oxide, 4,4'-Diaminophenyl ether, 4,4'-Oxybis(aniline), 4,4'-Oxydianiline, 4,4'-Oxydiphenylamine, 4-(4-Aminophenoxy)aniline, 4-(4-Aminophenoxy)phenylamine, 4-(4'-Aminophenoxy)benzenamine, 4-Aminophenyl ether, Bis(4-aminophenyl) ether, Bis(p-aminophenyl) ether, DDE, DPE, DPE (amine), Di(4-aminophenyl) ether, NSC 37075, NSC 6089, ODA, Oxybis(4-aminobenzene), Oxydi-p-phenylenediamine, Oxydianiline, p,p'-Diaminodiphenyl ether, p,p'-Oxybis[aniline], p,p'-Oxydianiline, p-Aminophenyl ether
InChI Formula InChI=1S/C12H12N2O/c13-9-1-5-11(6-2-9)15-12-7-3-10(14)4-8-12/h1-8H,13-14H2
SMILES Nc1ccc(Oc2ccc(N)cc2)cc1
IUPAC Name 4-(4-aminophenoxy)aniline
Molecular Weight (g/mol) 200.24
Formula Weight 200.095
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.