missing translation for 'onlineSavingsMsg'
Learn More

Bis(pentafluorophenyl) carbonate, 98+%

Product Code. 15401048
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15401048 5 g 5g
15491038 1 g 1g
15411048 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15401048 Supplier Thermo Scientific Alfa Aesar Supplier No. H54880.06
  • ONLINE EXCLUSIVE PRICE
    187.00€
    168.65€ / 5g
    Save 18.35€
    9% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 59483-84-0 | C13F10O3 | 394.12 g/mol

Bis(pentafluorophenyl) carbonate is a reagent used as a carbonyl equivalent and for various coupling reactions, especially for azapeptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(pentafluorophenyl) carbonate is a reagent used as a carbonyl equivalent and for various coupling reactions, especially for azapeptides.

Solubility
Soluble in chloroform, ethyl acetate and methylene chloride.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59483-84-0
Molecular Formula C13F10O3
Molecular Weight (g/mol) 394.12
MDL Number MFCD00368353
InChI Key IOVVFSGCNWQFQT-UHFFFAOYSA-N
Synonym bis pentafluorophenyl carbonate, bis perfluorophenyl carbonate, dipentafluorophenylcarbonate, pentafluorophenyl carbonate, di-pentafluorophenyl carbonate, bis 2,3,4,5,6-pentafluorophenyl carbonate, carbonic acid bis pentafluorophenyl ester, dpfpc, acmc-209mds
PubChem CID 2734833
IUPAC Name bis(2,3,4,5,6-pentafluorophenyl) carbonate
SMILES FC1=C(F)C(F)=C(OC(=O)OC2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F

Specifications

Melting Point 47°C to 50°C
Flash Point 110°C (230°F)
Linear Formula (C6F5O)2CO
Quantity 5 g
Beilstein 2029727
Solubility Information Soluble in chloroform,ethyl acetate and methylene chloride.
Formula Weight 394.12
Percent Purity ≥98%
Chemical Name or Material Bis(pentafluorophenyl) carbonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.