missing translation for 'onlineSavingsMsg'
Learn More

Bisoprolol Fumarate EP Impurity S, Mikromol™

Product Code. 17943026
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17943026 25 mg 25mg
17902066 100 mg 100mg
30460725 4 x 25 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17943026 Supplier Mikromol Supplier No. MM0460.140025
 Restricted Item
Estimated Shipment: 23-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Bisoprolol Fumarate EP Impurity S
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydroxybenzaldehyde
CAS 123-08-0
Analyte or Component Names 4-Hydroxybenzaldehyde
Shipping Condition Room Temperature
Molecular Formula C7 H6 O2
Certifications/Compliance ISO 17025
Synonym 4-Hydroxybenzaldehyde, Bisoprolol Fumarate Imp. S (EP), Bisoprolol Imp. S (EP), Bisoprolol Fumarate Impurity S, Bisoprolol Impurity S, p-Hydroxybenzaldehyde, 4-formylphenol, para-Hydroxybenzaldehyde
InChI Formula InChI=1S/C7H6O2/c8-5-6-1-3-7(9)4-2-6/h1-5,9H
SMILES Oc1ccc(C=O)cc1
IUPAC Name 4-hydroxybenzaldehyde
Molecular Weight (g/mol) 122.12
Quantity 25 mg
Formula Weight 122.0368 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.