missing translation for 'onlineSavingsMsg'
Learn More

Bisphenol A Monobenzyl Ether (>85%), TRC

Product Code. 30348643
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30348643 10 mg 10mg
30336105 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30348643 Supplier Toronto Research Chemicals Supplier No. TRCB51953510MG
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bisphenol A Monobenzyl Ether
CAS 42781-88-4
Molecular Formula C22H22O2
Quantity 10 mg
Synonym 4-[1-Methyl-1-[4-(phenylmethoxy)phenyl]ethyl]phenol
InChI Formula InChI=1S/C22H22O2/c1-22(2,18-8-12-20(23)13-9-18)19-10-14-21(15-11-19)24-16-17-6-4-3-5-7-17/h3-15,23H,16H2,1-2H3
SMILES CC(C)(C1=CC=C(C=C1)O)C2=CC=C(C=C2)OCC3=CC=CC=C3
IUPAC Name 4-[2-(4-phenylmethoxyphenyl)propan-2-yl]phenol
Molecular Weight (g/mol) 318.41
Formula Weight 318.162
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.