missing translation for 'onlineSavingsMsg'
Learn More

Bisphenol AP, TRC

Product Code. 30132435
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132435 25 g 25g
30133704 10 g 10g
30010155 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132435 Supplier Toronto Research Chemicals Supplier No. TRCB51948525G
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1571-75-1
Molecular Formula C20 H18 O2
Molecular Weight (g/mol) 290.36
Synonym Phenol, 4,4'-(α-methylbenzylidene)di- (6CI,7CI,8CI), 4,4'-(1-Phenylethylidene)bis[phenol], 1,1-Bis(4-hydroxyphenyl)-1-phenylethane, 1-Phenyl-1,1-bis(4-hydroxyphenyl)ethane, 4,4'-(1-Phenylethylidene)diphenol, 4,4'-(α-Methylbenzylidene)diphenol, Bis(4-hydroxyphenyl)methylphenylmethane, Bisp-Ap, Bisphenol ACP, Bisphenol AP, NK-AP, α,α-Bis(4-hydroxyphenyl)ethylbenzene
IUPAC Name 4-[1-(4-hydroxyphenyl)-1-phenylethyl]phenol
SMILES CC(c1ccccc1)(c2ccc(O)cc2)c3ccc(O)cc3

Specifications

Quantity 25 g
InChI Formula InChI=1S/C20H18O2/c1-20(15-5-3-2-4-6-15,16-7-11-18(21)12-8-16)17-9-13-19(22)14-10-17/h2-14,21-22H,1H3
Formula Weight 290.1307
Chemical Name or Material Bisphenol AP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.