missing translation for 'onlineSavingsMsg'
Learn More

Bisphenol S-monoisopropyl ether, Dr. Ehrenstorfer

Product Code. 30310363
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30310363 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30310363 Supplier Dr. Ehrenstorfer Supplier No. DREC10655947
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bisphenol S-monoisopropyl ether
CAS 95235-30-6
Analyte or Component Names Bisphenol S-monoisopropyl ether
Shipping Condition +20°C
Molecular Formula C15 H16 O4 S
Certifications/Compliance ISO 17034
Synonym 4-[[4-(1-Methylethoxy)phenyl]sulfonyl]phenol, 4-Hydroxy-4'-isopropoxy phenyl sulfone, 4-Hydroxy-4'-isopropoxydiphenyl sulfone, 4-Hydroxyphenyl 4-isopropoxyphenyl sulfone, 4-Isopropoxy-4'-hydroxydiphenyl sulfone, D 8, D 8 (sulfone), F 647, NYDS
InChI Formula InChI=1S/C15H16O4S/c1-11(2)19-13-5-9-15(10-6-13)20(17,18)14-7-3-12(16)4-8-14/h3-11,16H,1-2H3
SMILES CC(C)Oc1ccc(cc1)S(=O)(=O)c2ccc(O)cc2
IUPAC Name 4-(4-propan-2-yloxyphenyl)sulfonylphenol
Molecular Weight (g/mol) 292.35
Quantity 50 mg
Formula Weight 292.0769 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.