missing translation for 'onlineSavingsMsg'
Learn More

Bispyrazolone, Technical, Fisher Chemical

Product Code. 10570331 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10570331 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10570331 Supplier Fisher Chemical Supplier No. B/3620/44

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 7477-67-0
Melting Point 355°C
Color Cream
Packaging Plastic powder jar
Molecular Formula C20H18N4O2
MDL Number 20765
Quantity 5 g
InChI Key FORCWSNQDMPPOC-UHFFFAOYNA-N
SMILES CC1=NN(C(=O)C1C1C(C)=NN(C1=O)C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name 5,5'-dimethyl-2,2'-diphenyl-2H,2'H,3H,3'H,4H,4'H-[4,4'-bipyrazole]-3,3'-dione
Molecular Weight (g/mol) 346.39
Formula Weight 346.38g/mol
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.