Learn More
Bitolterol Mesylate, TRC
Product Code. 30180288
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180288 250 mg 250mg
30180264 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30180288 Supplier Toronto Research Chemicals Supplier No. TRCB591400250MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 30392-41-7
Molecular Formula C29H35NO8S
Molecular Weight (g/mol) 557.66
Synonym 4-Methyl-benzoic Acid 1,1'-[4-[2-[(1,1-Dimethylethyl)amino]-1-hydroxyethyl]-1,2-phenylene] Ester Methanesulfonate, 4-Methyl-benzoic Acid 4-[2-[(1,1-Dimethylethyl)amino]-1-hydroxyethyl]-1,2-phenylene Ester Methanesulfonate (salt), p-Toluic Acid 4-[2-(tert-Butylamino)-1-hydroxyethyl]-o-phenylene Ester Methanesulfonate (salt), (tert-Butyl)[β-hydroxy-3,4-bis(p-toluoyloxy)phenethyl]ammonium Methanesulfonate, Biterol, Bitoterol Methanesulfonate, Effectin, Tornalate, Win 32784
IUPAC Name [4-[2-(tert-butylamino)-1-hydroxyethyl]-2-(4-methylbenzoyl)oxyphenyl] 4-methylbenzoate;methanesulfonic acid
SMILES O=C(C1=CC=C(C)C=C1)OC2=CC(C(O)CNC(C)([C])C)=CC=C2OC(C3=CC=C(C)C=C3)=O.O=S(O)(C)=O
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C28H31NO5.CH4O3S/c1-18-6-10-20(11-7-18)26(31)33-24-15-14-22(23(30)17-29-28(3,4)5)16-25(24)34-27(32)21-12-8-19(2)9-13-21;1-5(2,3)4/h6-16,23,29-30H,17H2,1-5H3;1H3,(H,2,3,4)
Formula Weight 557.2083
Chemical Name or Material Bitolterol Mesylate