missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Blasticidin S Hydrochloride ReadyMade™ Solution

Product Code. 30044057
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 mL
10 x 1 mL
Unit Size:
10mL
1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30044057 10 x 1 mL 10mL
30044023 20 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30044057 Supplier TOKU-E™ Supplier No. B00610x1ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Blasticidin S Hydrochloride ReadyMade™ Solution is provided as a sterile solution of Blasticidin S Hydrochloride in HEPES buffer.

  • CAS Number: 3513-03-9
  • Molecular Formula: C17H26N8O5 · HCl
  • Molecular Weight: 458.90 g/mol
  • Storage Conditions: -20°C
  • Form: Liquid (clear, colorless/light yellow)
  • Source: Streptomyces griseochromogenes
  • Spectrum: Blasticidin S is active against mammalian and prokaryotic cells. Blasticidin S inhibits protein synthesis and is active against bacteria, fungi, nematodes, and tumor cells. The compound is used as a selection antibiotic for both eukaryotic and prokaryotic cells, and a marker for strain manipulation. It can also be used in genome editing using the CRISPR/Cas9 system
  • Mechanism of Action: Blasticidin S Hydrochloride inhibits protein synthesis in prokaryotic and eukaryotic cells by binding to the ribosomal P-site which strengthens tRNA binding and slows down and prevents subsequent peptide synthesis

Specifications

CAS 3-9-3513
Certifications/Compliance Purity >98.0%
Chemical Name or Material Blasticidin S
Color Colorless to light yellow
Content And Storage -20°C
Description Blasticidin S Hydrochloride ReadyMade™ Solution 10 mg/mL in 20 mM HEPES, pH 7.5
Concentration 9.7 to 10.3 mg/mL
Form Liquid
Product Type Antibiotic
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives HEPES
Without Additives None
Formulation Liquid, in HEPES buffer (pH 7.5)
For Use With (Application) Microbiology, Eukaryotic Cell Culture Applications
Product Line ReadyMade™ Solutions
Purity or Quality Grade >98.0%
Quantity 10 x 1 mL
Shipping Condition Dry Ice
Source Streptomyces griseochromogenes
Species Bacteria
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.