Learn More
BRAND™ BLAUBRAND™ Bulb Pipettes 1-Mark
Product Code. 16397967 Shop All Brand Products
Change view
Click to view available options
Volume (Metric):
1 mL
6 mL
20 mL
25 mL
Unit Size:
Pack of 12
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
16397967 1 mL Pack of 12
16347977 6 mL Pack of 6
16397977 20 mL Pack of 6
16307987 25 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16397967 Supplier BRAND™ Supplier No. 230602
  • ONLINE EXCLUSIVE PRICE
    101.00€
    77.00€ / Pack of 12
    Save 24.00€
    23% Off
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Class AS, 1-Mark Bulb Pipettes offer higher measurement accuracy than graduated pipettes.
  • AR-Glas™
  • Class 1-mark graduated
  • Includes batch certificate of conformity
TRUSTED_SUSTAINABILITY
Specifications
Class Class AS
Material AR-GLAS
Graduation Type 1-Mark Graduations
Includes Batch Certificate of Conformity
Volume (Metric) 1 mL
Type Bulb Pipet