missing translation for 'onlineSavingsMsg'
Learn More

Bleomycin A5 Hydrochloride Salt, TRC

Product Code. 30338474
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30338474 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30338474 Supplier Toronto Research Chemicals Supplier No. TRCB5957501MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bleomycin A5 hydrochloride
CAS 55658-47-4
Molecular Formula C57 H89 N19 O21 S2 . Cl H
Quantity 1 mg
Synonym N1-[3-[(4-Aminobutyl)amino]propyl]bleomycinamide Hydrochloride Salt
InChI Formula InChI=1S/C57H89N19O21S2.ClH/c1-22-35(73-48(76-46(22)61)27(14-33(60)80)68-15-26(59)47(62)86)52(90)75-37(43(28-16-65-21-69-28)95-56-45(41(84)39(82)31(17-77)94-56)96-55-42(85)44(97-57(63)92)40(83)32(18-78)93-55)53(91)70-24(3)38(81)23(2)49(87)74-36(25(4)79)51(89)67-13-8-34-71-30(20-98-34)54-72-29(19-99-54)50(88)66-12-7-11-64-10-6-5-9-58;/h16,19-21,23-27,31-32,36-45,55-56,64,68,77-79,81-85H,5-15,17-18,58-59H2,1-4H3,(H2,60,80)(H2,62,86)(H2,63,92)(H,65,69)(H,66,88)(H,67,89)(H,70,91)(H,74,87)(H,75,90)(H2,61,73,76);1H/t23-,24-,25-,26+,27+,31+,32-,36+,37+,38+,39-,40-,41+,42+,43+,44+,45+,55-,56+;/m1./s1
SMILES Cl.C[C@@H](O)[C@H](NC(=O)[C@H](C)[C@H](O)[C@@H](C)NC(=O)[C@@H](NC(=O)c1nc(nc(N)c1C)[C@H](CC(=O)N)NC[C@H](N)C(=O)N)[C@@H](O[C@@H]2O[C@@H](CO)[C@@H](O)[C@H](O)[C@@H]2O[C@H]3O[C@H](CO)[C@@H](O)[C@H](OC(=O)N)[C@@H]3O)c4c[nH]cn4)C(=O)NCCc5nc(cs5)c6nc(cs6)C(=O)NCCCNCCCCN
IUPAC Name [(2R,3S,4S,5R,6R)-2-[(2R,3S,4S,5S,6S)-2-[(1R,2S)-2-[[6-amino-2-[(1S)-3-amino-1-[[(2S)-2,3-diamino-3-oxopropyl]amino]-3-oxopropyl]-5-methylpyrimidine-4-carbonyl]amino]-3-[[(2R,3S,4R)-5-[[(2S,3R)-1-[2-[4-[4-[3-(4-aminobutylamino)propylcarbamoyl]-1,3-thiazol-2-yl]-1,3-thiazol-2-yl]ethylamino]-3-hydroxy-1-oxobutan-2-yl]amino]-3-hydroxy-4-methyl-5-oxopentan-2-yl]amino]-1-(1H-imidazol-4-yl)-3-oxopropoxy]-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl]oxy-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl] carbamate;hydrochloride
Molecular Weight (g/mol) 1477.02
Formula Weight 1475.5689
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.