Learn More
SCAT™ Blind Plugs, Green Line, PE
Product Code. 31454222 Shop All SCAT Europe Products
Change view
Click to view available options
Quantity:
10 Pk.
5 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31454222 10 Pk.
31454230 5 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 31454222 Supplier SCAT™ Supplier No. 460501
  • ONLINE EXCLUSIVE PRICE
    45.20€
    35.42€ / Pack of 1
    Save 9.78€
    21% Off
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
The Green Line Safety Caps stand for a conscious decision not to use PFAS, without compromising on quality and safety.
  • PFAS-free - future-proof and sustainable: The Green Line completely dispenses with PFAS and uses PEEK as a fluorine-free high-performance plastic that guarantees sustainability and future-proofing.
  • Full regulatory safety: Green Line products eliminate risks posed by upcoming PFAS bans and meet the highest environmental and ESG requirements.
  • Technical quality without compromise: The safety caps are 1:1 compatible with existing systems, chemically resistant, autoclavable and absolutely leak-proof in accordance with the SCAT standard.
  • Visibly sustainable: With their green coloring, the products make sustainability visible and demonstrate the laboratory's commitment to a PFAS-free future.
  • Development with foresight Green Line secures laboratory processes against regulatory risks in the long term and supports sustainable corporate strategies.
  • Product diversity and cost-effectiveness: The series offers a wide range of variants, starter kits and accessories for flexible, cost-effective and ready-to-use HPLC solutions.
  • In summary, SCAT Green Line stands for PFAS-free, regulatory safe, technically powerful and visibly sustainable laboratory solutions - without compromising on quality, tightness or compatibility. While conventional caps are based on PTFE, a representative of the so-called perpetual chemicals, the Green Line uses PEEK, a completely fluorine-free high-performance plastic with excellent chemical and thermal properties.
Specifications
Closure Color Colorless
Model Type 1
Quantity 10 Pk.
Size 1/4-28 UNF
For Use With (Application) PFAS-free analyses
For Use With (Equipment) Capillary connector
Product Type Blind Plug