missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Blue Sepharose™ High Performance

Product Code. 16032082 Shop All Cytiva Products
Change view
Click to view available options
Packaging Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Quantity unitSize
16032082 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16032082 Supplier Cytiva Supplier No. 90100123
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The carbohydrate nature of the agarose base provides a hydrophilic and chemically favorable environment for coupling. The highly crosslinked structure of the 34 μm spherical matrix ensures good chromatographic properties.

  • Blue Sepharose High Performance is a group specific adsorbent with affinity for a wide variety of enzymes
  • The dye ligand, Cibacron Blue F3G-A, is covalently attached to the matrix via the triazine part of the dye molecule
  • Some proteins interact biospecifically with the dye due to its structural similarity with nucleotide cofactors NAD+ and NADP+
  • Other proteins such as albumin and interferon, bind in a less specific manner by electrostatic and/or hydrophobic interactions with the aromatic anionic ligand
  • Dye ligands have greater versatility, which allows purification of several proteins without tedious preparation of a range of gels
  • The chemical stability of the adsorbent allows its continued re-use
  • Loss of activity through storage is not an issue
TRUSTED_SUSTAINABILITY

Specifications

Storage Requirements 4°C to 8°C
Particle Size 34 μm
Matrix Highly Cross-Linked, Spherical Agarose
pH Range 4 to 12
Storage Buffer 20% Ethanol
Ligand Type Cibacron™ Blue F3G-A
Packaging Quantity 50 mL
compliance-icons
Product Identifier
  • Blue Sepharose High Performance, 50 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.