missing translation for 'onlineSavingsMsg'
Learn More

Euro Protection™ Blue SPP Coverall

Product Code. 15852555
Change view
Click to view available options
Size:
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15852555 3X-Large Pack of 50
12033226 X-Large Pack of 50
12043226 2X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15852555 Supplier Euro Protection™ Supplier No. 42206
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Euro Protection™ Blue SPP Coverall is lightweight, disposable and features elastic at various points for good fit.

  • Hood with tightening elastic.
  • Zipper.
  • Tightening elastic on cuffs, waist, and ankles.
  • Light weight
  • Disposable
  • Comfortable

Specifications

Product Type General Purpose Coveralls
Size 3X-Large
Gender Unisex
Material Polypropylene
Color Blue
Disposable Single-use
Sterility Non-sterile
Resistance Abrasion
Certifications/Compliance (Europe) 1. Annexe II, Book III, Title I, Fourth Part of Code du Travail2. Article R412-233. EN530Article R4311-12
For Use With (Application) Visitors, Agrobusiness Industries, Cleaning, Packaging, Maintenance
Weight (Metric) 50 g/m2
Quantity 50/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.