missing translation for 'onlineSavingsMsg'
Learn More

Bmf Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication

Brand:  Novus Biologicals NBP1-76658-0.025mg

 View more versions of this product

Product Code. 18563263

  • 178.00€ / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Bmf Polyclonal specifically detects Bmf in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Bmf
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
Bcl2 modifying factor, bcl-2-modifying factor, FLJ00065
Rabbit
22 kDa
0.025 mg
Apoptosis, Cancer, Tumor Suppressors
90427
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Western Blot
1 mg/mL
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunofluorescence 5 μg/mL, Immunocytochemistry 10 μg/mL
NP_277038
BMF
Antibody was raised with a synthetic peptide corresponding to 15 amino acids near the amino terminus of human Bmf. The immunogen is located within the first 50 amino acids of Bmf. Amino Acid Squence: EPSQCVEELEDDV
Affinity Purified
RUO
Primary
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only