missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP-6 Antibody (SR1703), Novus Biologicals™
Rabbit Monoclonal Antibody
338.00€ - 449.00€
Specifications
| Antigen | BMP-6 |
|---|---|
| Clone | SR1703 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18637734
|
Novus Biologicals
NBP3-22013-100ul |
100 μg |
449.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18693734
|
Novus Biologicals
NBP3-22013-50ul |
50 μg |
338.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BMP-6 Monoclonal antibody specifically detects BMP-6 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
| BMP-6 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| BMP-6, bone morphogenetic protein 6, vegetal related growth factor (TGFB-related), vegetal-related (TGFB related) cytokine, VG-1-R, VG-1-related protein, Vg1-related sequence, VGR, VGR1, VGR-1 | |
| A synthesized peptide derived from human BMP-6 (Uniprot #: P22004) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1703 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cytokine Research | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 654 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title