Learn More
BMP1 Mouse anti-Human, Clone: OTI3E9, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505813S
Description
BMP1 Monoclonal antibody specifically detects BMP1 in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| BMP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P13497 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 649 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunocytochemistry, Western Blot | |
| OTI3E9 | |
| Unconjugated | |
| BMP1 | |
| BMP; Bmp1; BMP-1; Bone morphogenetic protein; Bone morphogenetic protein 1; FLJ44432; mammalian tolloid protein; mTld; OI13; PCOLC; Pcp; PCP2; pCP-2; procollagen C-endopeptidase; procollagen C-proteinase; procollagen C-proteinase 3; TLD | |
| Human recombinant protein fragment corresponding to amino acids 225-617 of human BMP1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.