missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP9 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17769-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. Studies in rodents suggest that this protein plays a role in the adult liver and in differentiation of cholinergic central nervous system neurons.Specifications
| BMP9 | |
| Polyclonal | |
| Unconjugated | |
| GDF2 | |
| BMP 9, BMP9, Bone morphogenetic protein 9, GDF 2, GDF2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2658 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9UK05 | |
| GDF2 | |
| BMP9 Fusion Protein Ag12177 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction