missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BMPER Monoclonal Antibody (349920)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561134
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene encodes a secreted protein that interacts with, and inhibits bone morphogenetic protein (BMP) function. It has been shown to inhibit BMP2- and BMP4-dependent osteoblast differentiation and BMP-dependent differentiation of the chondrogenic cells. Mutations in this gene are associated with a lethal skeletal disorder, diaphanospondylodysostosis.
Specifications
| BMPER | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q8CJ69 | |
| Bmper | |
| Mouse myeloma cell line NS0-derived recombinant mouse Crossveinless-2/CV-2, Val34-Arg685. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 349920 | |
| Unconjugated | |
| Bmper | |
| BMP-Binding Endothelial Regulator Protein; CRIM3 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 73230 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction