missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMPR-IB/ALK-6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-14889
This item is not returnable.
View return policy
Description
BMPR-IB/ALK-6 Polyclonal specifically detects BMPR-IB/ALK-6 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| BMPR-IB/ALK-6 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0) and 20% Glycerol with 0.025% Proclin 300 | |
| activin receptor-like kinase 6, ALK-6, ALK6CDw293, BMP type-1B receptor, BMPR-1B, bone morphogenetic protein receptor type-1B, bone morphogenetic protein receptor, type IB, CDw293 antigen, EC 2.7.11, EC 2.7.11.30, serine/threonine receptor kinase | |
| Rabbit | |
| 57 kDa | |
| 0.1 mL | |
| Mesenchymal Stem Cell Markers, Protein Kinase, Stem Cell Markers | |
| 658 | |
| Human, Mouse, Rat, Bovine | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 0.81mg/mL | |
| Western Blot 1:500-1:10000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen 1:100-1:1000 | |
| O00238 | |
| BMPR1B | |
| Recombinant protein encompassing a sequence within the center region of human BMPR1B. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Predicted cross-reactivity based on sequence identity: Xenopus tropicalis (94%) and Bovine (99%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction