Learn More
BMT2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21744-1-AP-150UL
Description
The BMT2 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human BMT2. This antibody recognizes human, mouse, rat antigen. The BMT2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| BMT2 | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat |
| Western Blot | |
| Unconjugated | |
| 150 μL | |
| 154743 | |
| IgG |