missing translation for 'onlineSavingsMsg'
Learn More

BOC-D-Aspartic acid beta-benzyl ester, 98%, Thermo Scientific™

Product Code. 11985571 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 11985571

Brand: Thermo Scientific Acros 442870050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 51186-58-4
Melting Point 101°C to 105°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Molecular Formula C16H21NO6
Quantity 5 g
Specific Rotation 19.56
Synonym boc-d-asp obzl-oh, boc-d-aspartic acid 4-benzyl ester, r-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, 2r-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-alpha-t-boc-d-aspartic acid beta-benzyl ester, boc-d-asparticacid4-benzylester, pubchem11179, boc-d-asp obn-oh, boc-d-aspartic acid b-benzyl ester, boc-d-aspartic acid ?-benzyl ester
InChI Key SOHLZANWVLCPHK-GFCCVEGCSA-N
SMILES CC(C)(C)OC(=O)NC(CC(=O)OCC1=CC=CC=C1)C(=O)O
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid
Molecular Weight (g/mol) 323.35
PubChem CID 6994663
Formula Weight 323.35
Percent Purity 98%
Physical Form Powder
Chemical Name or Material BOC-D-Aspartic acid β-benzyl ester
Show More Show Less
Product Identifier
  • BOC-D-Aspartic acid beta-benzyl ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.