Learn More
Invitrogen™ BODIPY™ FL (4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Propionic Acid)
Product Code. 11590246
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11590246 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11590246 Supplier Invitrogen™ Supplier No. D2183
BODIPY™ FL (4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Propionic Acid)
The amine-reactive BODIPY™ FL propionic acid can be used to create green-fluorescent bioconjugates following carbodiimide-activation of the carboxylic acid on the dye and reaction with amines. The electronically neutral BODIPY™ FL dye has the most fluorescein-like spectra of the green-fluorescent BODIPY™ dyes.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5 mg
Product Type FL Propionic Acid
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Carboxylic Acid
Label or Dye BODIPY™ FL
Label Type BODIPY Dyes
Product Line BODIPY