Learn More
Bohlender™ BOLA™ Filtering Discs
Filtering discs made of porous PTFE.
Brand:  Bohlender™ N1565-42
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 17117514
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • Made of porous PTFE
  • Thickness: 1.0 mm
Specifications
Specifications
PTFE
6362 mm2
0.04 in.
90 mm
1 μm
10 Pk.
1 mm