Learn More
Bohlender™ BOLA™ Filtering Discs
Filtering discs made of porous PTFE.
Brand: Bohlender™ N1565-06
Additional Details : Weight : 0.27850kg
Description
- Made of porous PTFE
- Thickness: 1.0 mm
Specifications
PTFE | |
1735 mm2 | |
0.04 in. | |
47 mm |
1 μm | |
10 Pk. | |
1 mm |