missing translation for 'onlineSavingsMsg'
Learn More

Bohlender™ BOLA™ Filtering Discs

Filtering discs made of porous PTFE.

Brand:  Bohlender™ N1565-18

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 17197504

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »


Description

Description

  • Made of porous PTFE
  • Thickness: 1.0 mm
Specifications

Specifications

PTFE
1735 mm2
0.04 in.
47 mm
10 μm
10 Pk.
1 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers