Learn More
Bohlender™ BOLA™ Filtering Discs
Filtering discs made of porous PTFE.
Brand:  Bohlender™ N1565-24
Additional Details : Weight : 0.19000kg
 View more versions of this product
Product Code. 17814024
  • 94.98€ / Pack of 10
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
Description
Description
  • Made of porous PTFE
  • Thickness: 1.0 mm
Specifications
Specifications
PTFE
1735 mm2
0.04 in.
47 mm
25 μm
10 Pk.
1 mm