missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid 1% Solution and Indicator For Kjeldahl N Det., Chem Lab

Product Code. 11989162
Change view
Click to view available options
Quantity:
5 L
10 L
Unit Size:
10 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11989162 10 L 10 L
11979162 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11989162 Supplier Chem-Lab Supplier No. CL02.0204.9010
  • ONLINE EXCLUSIVE PRICE
    97.00€
    51.30€ / 10 L
    Save 45.70€
    47% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Color Gray-Green
Melting Point 0°C
Boiling Point 100°C
Density 1 g/mol
Packaging Polyethylene
Quantity 10 L
Physical Form Liquid
Molecular Weight (g/mol) 61.83
compliance-icons
Product Identifier
  • Boric acid 1% solution + indicator
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Boric acid, Methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.