missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, ACS Reagent Grade, Ricca Chemical

Product Code. 31750694
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Unit Size:
1000g
2.50 L
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31750694 2.5 kg 2.50 L
31751134 500 g 500g
31751105 1 kg 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31750694 Supplier Ricca Chemical Company Supplier No. RDCB05002.5A5
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
IUPAC Name boric acid
SMILES OB(O)O

Specifications

CAS Max % 100
Heavy Metals (as Pb) max. 0.001%
Non_volatile Matter max. 0.05%
Packaging Natural Poly Bottle
Quantity 2.5 kg
Percent Purity min 99.5%
Grade ACS
Chemical Name or Material Boric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.