missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid MP Biomedicals

Product Code. 11460932 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460932 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11460932 Supplier MP Biomedicals Supplier No. 0219507401
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 10043-35-3
Density 1.5172 g/cm3 at 14 °C
Quantity 1 kg
Molecular Formula BH3O3
MDL Number MFCD00011337
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
SMILES OB(O)O
IUPAC Name boric acid
Molecular Weight (g/mol) 61.83
PubChem CID 7628
ChEBI CHEBI:33118
Formula Weight 61.83
Chemical Name or Material Boric Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.