missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, N.F., A.C.S., J.T. Baker™

Product Code. 15528454 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15528454 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15528454 Supplier Avantor J.T.Baker Supplier No. 790501

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 10043-35-3
Packaging Poly Bottle
Specific Gravity 1L = 1.44kg
Quantity 500 g
Molecular Formula BH3O3
MDL Number MFCD00011337
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
SMILES OB(O)O
IUPAC Name boric acid
Molecular Weight (g/mol) 61.83
PubChem CID 7628
ChEBI CHEBI:33118
Formula Weight 61.83
Grade ACS/NF
Chemical Name or Material Boric Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.