missing translation for 'onlineSavingsMsg'
Learn More

Borneol, TRC

Product Code. 30133925
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133925 25 g 25g
30009291 5 g 5g
17970835 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133925 Supplier Toronto Research Chemicals Supplier No. TRCB67542025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 507-70-0
Molecular Formula C10 H18 O
Molecular Weight (g/mol) 154.25
Synonym Bicyclo[2.2.1]heptan-2-ol, 1,7,7-trimethyl-, (1R,2S,4R)-rel-, Bicyclo[2.2.1]heptan-2-ol, 1,7,7-trimethyl-, endo-, Borneol (8CI), (±)-Borneol, 2-Borneol, 2-endo-Bornyl alcohol, Camphol, NSC 60223, dl-Borneol, endo-(±)-Bornan-2-ol, endo-2-Hydroxy-1,7,7-trimethylnorbornane, endo-Borneol, rel-(1R,2S,4R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol, rel-(1S,2R,4S)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol
IUPAC Name (1S,3R,4S)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-ol
SMILES CC1(C)[C@H]2CC[C@]1(C)[C@H](O)C2

Specifications

Quantity 25 g
InChI Formula InChI=1S/C10H18O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7-8,11H,4-6H2,1-3H3/t7-,8+,10+/m0/s1
Formula Weight 154.1358
Percent Purity >95%
Chemical Name or Material Borneol
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.