missing translation for 'onlineSavingsMsg'
Learn More

Boron trifluoride tetrahydrofuran complex 50%, Thermo Scientific™

Product Code. 10753134 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
500mL; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10753134 500mL; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10753134 Supplier Thermo Scientific Acros Supplier No. 388635000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Lewis acid
TRUSTED_SUSTAINABILITY

Specifications

CAS 462-34-0
Quantity 500mL
Assay Percent Range 50%
Molecular Formula C4H8BF3O
MDL Number MFCD00040372
Synonym boron trifluoride tetrahydrofuran complex, trifluoro tetrahydro-1h-furan-1-ium-1-yl borate, pubchem7062, trifluoro oxolan-1-ium-1-yl boranuide, borontrifluoride tetrahydrofuran complex, boron trifluoride tetrahydrofuran complex, contains sulfur dioxide as peroxide formation suppressor
InChI Key CHNLPLHJUPMEOI-UHFFFAOYSA-N
SMILES FB(F)F.C1CCOC1
IUPAC Name oxolane; trifluoroborane
Molecular Weight (g/mol) 139.91
PubChem CID 60146129
Formula Weight 139.91
Percent Purity 48 to 50%
Identification Confirmed
Packaging Glass bottle
Density 1.2700g/mL
Chemical Name or Material Boron trifluoride tetrahydrofuran complex
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.