Learn More
Witeg™ Borosilicate glass 3.3 Round bottomed flasks
Specifications
Specifications
| Certifications/Compliance | DIN 12348 |
| Capacity (Metric) | 1000 mL |
| Material | Borosilicate Glass (3.3) |
| Neck Style | Center |
| Bottom Shape | Round |