Learn More
BRAND™ Borosilicate Glass Desiccator
Product Code. 10203731 Shop All Brand Products
Change view
Click to view available options
Diameter (Metric) Outer:
215 mm
270 mm
320 mm
Height (Metric):
316 mm
368 mm
417 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Height (Metric) Unit Size
10203731 270 mm 368 mm Each
10738051 215 mm 316 mm Each
10514262 320 mm 417 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10203731 Supplier BRAND™ Supplier No. 65238
Estimated Shipment: 18-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Manufactured from borosilicate glass complies with DIN 12491 standards. BRAND™ Borosilicate Glass Desiccator has precisely ground flat flanges. Base ring surface is studded to improve stability and help prevent cracks. Supplied with 24/29 socket and interchangeable borosilicate glass stopcock.
  • Manufactured from borosilicate glass.
  • Complies with DIN 12491 standards.
  • Precisely ground flat flanges.
  • Base ring surface studded to improve stability and help prevent cracks.
  • Interchangeable lid.
  • Supplied with 24/29 socket and interchangeable borosilicate glass stopcock.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Outer 270 mm
Material Borosilicate Glass
Height (Metric) 368 mm
Includes Socket in the lid with stopcock
Product Type Desiccator
Quantity 1