Description
- Manufactured from borosilicate glass.
- Complies with DIN 12491 standards.
- Precisely ground flat flanges.
- Base ring surface studded to improve stability and help prevent cracks.
- Interchangeable lid.
- Supplied with 24/29 socket and interchangeable borosilicate glass stopcock.
Specifications
Specifications
| Diameter (Metric) Outer | 320 mm |
| Description | Precisely ground flat flanges. Base ring surface studded to improve stability and help prevent cracks. |
| Material | Borosilicate Glass |
| Height (Metric) | 417 mm |
| Includes | Socket in the lid with stopcock |
| Color | Clear |
| Quantity | 1 |