Learn More
Cytiva Borosilicate Glass Ethylene Propylene Complete Superloop
Product Code. 11535704 Shop All Cytiva Products
Change view
Click to view available options
Volume (Metric):
150 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11535704 150 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11535704 Supplier Cytiva Supplier No. 18102385
Estimated Shipment: 01-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Use with aqueous solutions and alcohols. Cytiva Lifescience™ Borosilicate Glass Ethylene Propylene Complete Superloop contains a springloaded valve made from titanium.
  • Glass tube
  • Graduated scale of 5mL increments and seals against the inner end pieces.
  • A springloaded valve made from titanium is located in the centre of the movable seal
  • Tubing and connectors have to be ordered separately.
TRUSTED_SUSTAINABILITY
Specifications
Size (Metric) 45 mm dia. x 340 mm L
For Use With (Application) Aqueous Solutions and Alcohols
Max. Pressure 1 bar
Length (Metric) 340 mm
Diameter (Metric) Inner 45 mm
Volume (Metric) 150 mL