Learn More
Fisherbrand™ Glass Rectangular Coverslips
Product Code. 12393128 Shop All Fisherbrand Products
Change view
Click to view available options
Thickness (Metric):
#1 (0.13 to 0.16 mm)
#1.5 (0.16 to 0.19 mm)
Length (Metric):
22 mm
24 mm
Width (Metric):
26 mm
32 mm
40 mm
50 mm
Unit Size:
Pack of 100
Pack of 200
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thickness (Metric) Length (Metric) Width (Metric) unitSize
12393128 #1 (0.13 to 0.16 mm) 24 mm 32 mm Pack of 100
12343128 #1 (0.13 to 0.16 mm) 22 mm 26 mm Pack of 200
12353128 #1 (0.13 to 0.16 mm) 22 mm 32 mm Pack of 100
12363128 #1 (0.13 to 0.16 mm) 22 mm 40 mm Pack of 100
12373128 #1 (0.13 to 0.16 mm) 22 mm 50 mm Pack of 100
12383138 #1.5 (0.16 to 0.19 mm) 22 mm 50 mm Pack of 100
11348503 #1 (0.13 to 0.16 mm) 24 mm 40 mm Pack of 100
12303148 #1.5 (0.16 to 0.19 mm) 24 mm 50 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 12393128
  • ONLINE EXCLUSIVE PRICE
    3.81€
    2.74€ / Pack of 100
    Save 1.07€
    28% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Glass Rectangular Coverslips are available in a variety of sizes. Packed in plastic boxes to prevent breakage.
  • Manufactured from clear soda glass
  • Consistent in cut, thickness and dimensional accuracy
  • Free from blisters and flaws
Specifications
Product Type Coverslip
Shape Rectangle
Material Soda Lime Glass
Length (Metric) 24 mm
Width (Metric) 32 mm
Thickness (Metric) #1 (0.13 to 0.16 mm)
Quantity 100 Pk.